Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12192756-12193250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFRSF1B
Entrez Gene ID
7133
Gene Name
TNF receptor superfamily member 1B
Gene Aliases
CD120b, TBPII, TNF-R-II, TNF-R75, TNFBR, TNFR1B, TNFR2, TNFR80, p75, p75TNFR
UniGene
Hs.256278
Gene Chromosome Location
Chr.1: 12166948-12209222 on Build GRCh38
Transcripts
NM_001066.2 XM_011542060.2 XM_011542063.2 XM_017002211.1 XM_017002212.1 XM_017002213.1 XM_017002214.1 XM_017002215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
146406 c.587T>G p.M196R Chr.1: 12192898-12192898 on Build GRCh38
3747515 c.694G>A p.E232K Chr.1: 12193005-12193005 on Build GRCh38
1600661 c.641T>C p.V214A Chr.1: 12192952-12192952 on Build GRCh38
5377435 c.719C>T p.S240F Chr.1: 12193030-12193030 on Build GRCh38
3801492 c.643C>T p.H215Y Chr.1: 12192954-12192954 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886628953 Chr.1: 12192809-12192809 on Build GRCh38
rs952652900 Chr.1: 12192852-12192852 on Build GRCh38
rs199873050 Chr.1: 12192919-12192919 on Build GRCh38
rs757216527 Chr.1: 12193205-12193205 on Build GRCh38
rs778888553 Chr.1: 12193095-12193095 on Build GRCh38
rs771900114 Chr.1: 12192992-12192992 on Build GRCh38
rs140857317 Chr.1: 12192930-12192930 on Build GRCh38
rs770553406 Chr.1: 12193156-12193159 on Build GRCh38
rs778608285 Chr.1: 12192846-12192847 on Build GRCh38
rs753843207 Chr.1: 12193169-12193169 on Build GRCh38
rs200254914 Chr.1: 12192876-12192876 on Build GRCh38
rs867083487 Chr.1: 12192841-12192841 on Build GRCh38
rs1061622 Chr.1: 12192898-12192898 on Build GRCh38
rs1035218772 Chr.1: 12192913-12192913 on Build GRCh38
rs768027709 Chr.1: 12192899-12192899 on Build GRCh38
rs753626242 Chr.1: 12193192-12193192 on Build GRCh38
rs146983380 Chr.1: 12193079-12193079 on Build GRCh38
rs754426669 Chr.1: 12192821-12192821 on Build GRCh38
rs537233011 Chr.1: 12192884-12192884 on Build GRCh38
rs750651159 Chr.1: 12193162-12193162 on Build GRCh38
rs746585421 Chr.1: 12193130-12193132 on Build GRCh38
rs981800406 Chr.1: 12192904-12192904 on Build GRCh38
rs773695049 Chr.1: 12192987-12192987 on Build GRCh38
rs888291217 Chr.1: 12193055-12193055 on Build GRCh38
rs554329036 Chr.1: 12192822-12192822 on Build GRCh38
rs150329910 Chr.1: 12193027-12193027 on Build GRCh38
rs941728356 Chr.1: 12193213-12193213 on Build GRCh38
rs923246282 Chr.1: 12192807-12192807 on Build GRCh38
rs536867737 Chr.1: 12192869-12192869 on Build GRCh38
rs559822776 Chr.1: 12192789-12192789 on Build GRCh38
rs761941588 Chr.1: 12193149-12193149 on Build GRCh38
rs765420868 Chr.1: 12192823-12192823 on Build GRCh38
rs1057504327 Chr.1: 12192980-12192980 on Build GRCh38
rs772890535 Chr.1: 12193095-12193096 on Build GRCh38
rs759635294 Chr.1: 12192793-12192793 on Build GRCh38
rs763535034 Chr.1: 12193140-12193140 on Build GRCh38
rs564617314 Chr.1: 12193080-12193080 on Build GRCh38
rs745814265 Chr.1: 12192871-12192871 on Build GRCh38
rs764190618 Chr.1: 12192991-12192991 on Build GRCh38
rs923160179 Chr.1: 12193192-12193193 on Build GRCh38
rs771703889 Chr.1: 12193102-12193102 on Build GRCh38
rs750633484 Chr.1: 12193065-12193065 on Build GRCh38
rs551866527 Chr.1: 12193105-12193105 on Build GRCh38
rs780026833 Chr.1: 12192975-12192975 on Build GRCh38
rs771672384 Chr.1: 12192870-12192872 on Build GRCh38
rs775398223 Chr.1: 12192882-12192882 on Build GRCh38
rs757100808 Chr.1: 12192865-12192865 on Build GRCh38
rs201545665 Chr.1: 12192924-12192924 on Build GRCh38
rs764261095 Chr.1: 12193028-12193028 on Build GRCh38
rs267597960 Chr.1: 12193031-12193031 on Build GRCh38
rs764019731 Chr.1: 12192944-12192944 on Build GRCh38
rs770267357 Chr.1: 12193012-12193012 on Build GRCh38
rs774921744 Chr.1: 12193143-12193143 on Build GRCh38
rs746599817 Chr.1: 12193106-12193106 on Build GRCh38
rs761853712 Chr.1: 12193052-12193052 on Build GRCh38
rs906035722 Chr.1: 12192896-12192896 on Build GRCh38
rs760848007 Chr.1: 12193164-12193164 on Build GRCh38
rs2228494 Chr.1: 12192870-12192870 on Build GRCh38
rs976411216 Chr.1: 12192794-12192794 on Build GRCh38
rs983094931 Chr.1: 12193195-12193195 on Build GRCh38
rs761331191 Chr.1: 12192920-12192920 on Build GRCh38
rs757378017 Chr.1: 12192959-12192959 on Build GRCh38
rs747930689 Chr.1: 12192847-12192849 on Build GRCh38
rs1768642 Chr.1: 12192835-12192835 on Build GRCh38
rs772968093 Chr.1: 12193001-12193001 on Build GRCh38
rs767529430 Chr.1: 12193153-12193153 on Build GRCh38
rs777077445 Chr.1: 12192925-12192925 on Build GRCh38
rs767116063 Chr.1: 12193183-12193184 on Build GRCh38
rs749124537 Chr.1: 12192979-12192979 on Build GRCh38
rs775673222 Chr.1: 12193130-12193130 on Build GRCh38
rs778099535 Chr.1: 12192851-12192851 on Build GRCh38
rs199974127 Chr.1: 12193086-12193086 on Build GRCh38
rs745706109 Chr.1: 12192970-12192970 on Build GRCh38
rs755918822 Chr.1: 12193073-12193073 on Build GRCh38
rs1047110705 Chr.1: 12193045-12193045 on Build GRCh38
rs950690531 Chr.1: 12193194-12193194 on Build GRCh38
rs766601906 Chr.1: 12192817-12192817 on Build GRCh38
rs557507770 Chr.1: 12193107-12193107 on Build GRCh38
rs1047451026 Chr.1: 12192831-12192831 on Build GRCh38
rs5746027 Chr.1: 12193017-12193017 on Build GRCh38
rs964618029 Chr.1: 12192792-12192792 on Build GRCh38
rs549564583 Chr.1: 12192931-12192931 on Build GRCh38
rs754770781 Chr.1: 12193198-12193198 on Build GRCh38
rs145736387 Chr.1: 12192982-12192982 on Build GRCh38
rs1005304435 Chr.1: 12193062-12193062 on Build GRCh38
rs1024014210 Chr.1: 12193075-12193075 on Build GRCh38
rs375948624 Chr.1: 12193044-12193044 on Build GRCh38
rs5746026 Chr.1: 12193005-12193005 on Build GRCh38
rs373965812 Chr.1: 12192846-12192846 on Build GRCh38
rs759966102 Chr.1: 12192926-12192926 on Build GRCh38
rs767797172 Chr.1: 12193060-12193060 on Build GRCh38
rs751807806 Chr.1: 12192945-12192945 on Build GRCh38
rs781375513 Chr.1: 12192965-12192965 on Build GRCh38
rs747179551 Chr.1: 12192859-12192859 on Build GRCh38
rs758561163 Chr.1: 12192933-12192933 on Build GRCh38
rs764976691 Chr.1: 12193211-12193211 on Build GRCh38

View Full Product Details