Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 27924455-27924962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDX1
Entrez Gene ID
3651
Gene Name
pancreatic and duodenal homeobox 1
Gene Aliases
GSF, IDX-1, IPF1, IUF1, MODY4, PAGEN1, PDX-1, STF-1
UniGene
Hs.32938
Gene Chromosome Location
Chr.13: 27919982-27926314 on Build GRCh38
Transcripts
NM_000209.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4937202 c.646G>A p.G216R Chr.13: 27924495-27924495 on Build GRCh38
6989392 c.822C>T p.V274V Chr.13: 27924671-27924671 on Build GRCh38
6852464 c.848G>A p.R283Q Chr.13: 27924697-27924697 on Build GRCh38
6856534 c.714_716delGCC p.P244delP Chr.13: 27924563-27924565 on Build GRCh38
6743489 c.615G>A p.E205E Chr.13: 27924464-27924464 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751645380 Chr.13: 27924549-27924549 on Build GRCh38
rs778200349 Chr.13: 27924857-27924857 on Build GRCh38
rs551863908 Chr.13: 27924885-27924885 on Build GRCh38
rs193922357 Chr.13: 27924575-27924576 on Build GRCh38
rs748376235 Chr.13: 27924500-27924501 on Build GRCh38
rs774759417 Chr.13: 27924729-27924729 on Build GRCh38
rs201343646 Chr.13: 27924508-27924508 on Build GRCh38
rs753650635 Chr.13: 27924509-27924509 on Build GRCh38
rs749996964 Chr.13: 27924483-27924483 on Build GRCh38
rs901732228 Chr.13: 27924778-27924778 on Build GRCh38
rs530404627 Chr.13: 27924739-27924739 on Build GRCh38
rs955359808 Chr.13: 27924908-27924908 on Build GRCh38
rs753881947 Chr.13: 27924568-27924568 on Build GRCh38
rs770346335 Chr.13: 27924641-27924641 on Build GRCh38
rs1040058277 Chr.13: 27924742-27924742 on Build GRCh38
rs950269577 Chr.13: 27924704-27924704 on Build GRCh38
rs764011596 Chr.13: 27924494-27924494 on Build GRCh38
rs762020686 Chr.13: 27924563-27924563 on Build GRCh38
rs1031073225 Chr.13: 27924829-27924829 on Build GRCh38
rs367625089 Chr.13: 27924543-27924543 on Build GRCh38
rs996109320 Chr.13: 27924796-27924796 on Build GRCh38
rs771764758 Chr.13: 27924518-27924518 on Build GRCh38
rs1017467511 Chr.13: 27924552-27924552 on Build GRCh38
rs569303756 Chr.13: 27924806-27924806 on Build GRCh38
rs899889574 Chr.13: 27924850-27924850 on Build GRCh38
rs55957037 Chr.13: 27924918-27924918 on Build GRCh38
rs776932903 Chr.13: 27924482-27924482 on Build GRCh38
rs754867917 Chr.13: 27924512-27924512 on Build GRCh38
rs768725073 Chr.13: 27924720-27924720 on Build GRCh38
rs956678834 Chr.13: 27924580-27924580 on Build GRCh38
rs150559931 Chr.13: 27924510-27924510 on Build GRCh38
rs137852787 Chr.13: 27924519-27924519 on Build GRCh38
rs17847223 Chr.13: 27924789-27924789 on Build GRCh38
rs1056035961 Chr.13: 27924688-27924688 on Build GRCh38
rs1001135445 Chr.13: 27924826-27924826 on Build GRCh38
rs940344638 Chr.13: 27924709-27924709 on Build GRCh38
rs763082291 Chr.13: 27924491-27924491 on Build GRCh38
rs200411304 Chr.13: 27924542-27924542 on Build GRCh38
rs955246004 Chr.13: 27924576-27924576 on Build GRCh38
rs756067225 Chr.13: 27924504-27924504 on Build GRCh38
rs781657590 Chr.13: 27924608-27924608 on Build GRCh38
rs920108298 Chr.13: 27924619-27924619 on Build GRCh38
rs374397203 Chr.13: 27924769-27924769 on Build GRCh38
rs930365498 Chr.13: 27924669-27924669 on Build GRCh38
rs930419631 Chr.13: 27924484-27924484 on Build GRCh38
rs981314993 Chr.13: 27924609-27924609 on Build GRCh38
rs372765489 Chr.13: 27924528-27924528 on Build GRCh38
rs937660726 Chr.13: 27924686-27924686 on Build GRCh38
rs751672722 Chr.13: 27924498-27924498 on Build GRCh38
rs777512281 Chr.13: 27924581-27924581 on Build GRCh38
rs532113600 Chr.13: 27924811-27924811 on Build GRCh38
rs996947186 Chr.13: 27924853-27924853 on Build GRCh38
rs199644078 Chr.13: 27924565-27924565 on Build GRCh38
rs561437933 Chr.13: 27924710-27924710 on Build GRCh38
rs754818945 Chr.13: 27924572-27924572 on Build GRCh38
rs752553392 Chr.13: 27924573-27924573 on Build GRCh38
rs368507502 Chr.13: 27924513-27924513 on Build GRCh38
rs527882800 Chr.13: 27924635-27924635 on Build GRCh38
rs371525768 Chr.13: 27924546-27924546 on Build GRCh38
rs541584830 Chr.13: 27924660-27924660 on Build GRCh38
rs759874752 Chr.13: 27924539-27924539 on Build GRCh38
rs775667083 Chr.13: 27924666-27924666 on Build GRCh38
rs370787126 Chr.13: 27924606-27924606 on Build GRCh38
rs756811291 Chr.13: 27924584-27924584 on Build GRCh38
rs887273572 Chr.13: 27924755-27924755 on Build GRCh38
rs918169516 Chr.13: 27924592-27924592 on Build GRCh38
rs907660995 Chr.13: 27924706-27924706 on Build GRCh38
rs1029635032 Chr.13: 27924880-27924880 on Build GRCh38
rs1049166702 Chr.13: 27924495-27924495 on Build GRCh38
rs777164146 Chr.13: 27924530-27924530 on Build GRCh38
rs956814094 Chr.13: 27924842-27924842 on Build GRCh38
rs746492280 Chr.13: 27924583-27924583 on Build GRCh38
rs754817283 Chr.13: 27924697-27924697 on Build GRCh38
rs193922359 Chr.13: 27924577-27924578 on Build GRCh38
rs765415673 Chr.13: 27924485-27924485 on Build GRCh38
rs926396364 Chr.13: 27924677-27924677 on Build GRCh38
rs747825143 Chr.13: 27924516-27924516 on Build GRCh38
rs969702158 Chr.13: 27924607-27924607 on Build GRCh38
rs193922360 Chr.13: 27924622-27924622 on Build GRCh38
rs193922358 Chr.13: 27924574-27924574 on Build GRCh38
rs776610329 Chr.13: 27924500-27924500 on Build GRCh38
rs746545394 Chr.13: 27924524-27924524 on Build GRCh38

View Full Product Details