Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 31246691-31247193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GLCT
Entrez Gene ID
145173
Gene Name
beta 3-glucosyltransferase
Gene Aliases
B3GALTL, B3GTL, B3Glc-T, Gal-T, beta3Glc-T
UniGene
Hs.13205
Gene Chromosome Location
Chr.13: 31199975-31332276 on Build GRCh38
Transcripts
NM_194318.3 XM_006719768.3 XM_011534936.1 XM_011534938.2 XM_017020395.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5527220 c.286C>T p.L96F Chr.13: 31247038-31247038 on Build GRCh38
4488555 c.334C>G p.P112A Chr.13: 31247086-31247086 on Build GRCh38
4046988 c.322T>C p.W108R Chr.13: 31247074-31247074 on Build GRCh38
6138801 c.288C>G p.L96L Chr.13: 31247040-31247040 on Build GRCh38
1245872 c.324G>T p.W108C Chr.13: 31247076-31247076 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1051718817 Chr.13: 31246959-31246959 on Build GRCh38
rs903495992 Chr.13: 31247132-31247132 on Build GRCh38
rs367837499 Chr.13: 31246968-31246968 on Build GRCh38
rs770487360 Chr.13: 31247073-31247073 on Build GRCh38
rs932935933 Chr.13: 31247043-31247043 on Build GRCh38
rs372324692 Chr.13: 31247087-31247087 on Build GRCh38
rs998810767 Chr.13: 31246783-31246783 on Build GRCh38
rs80338850 Chr.13: 31247104-31247104 on Build GRCh38
rs556532817 Chr.13: 31246952-31246953 on Build GRCh38
rs186910617 Chr.13: 31247155-31247155 on Build GRCh38
rs370425638 Chr.13: 31247025-31247025 on Build GRCh38
rs567637098 Chr.13: 31246952-31246952 on Build GRCh38
rs200376294 Chr.13: 31247067-31247067 on Build GRCh38
rs752416772 Chr.13: 31246982-31246982 on Build GRCh38
rs140139669 Chr.13: 31246891-31246891 on Build GRCh38
rs112738167 Chr.13: 31246814-31246814 on Build GRCh38
rs371768901 Chr.13: 31246951-31246951 on Build GRCh38
rs370517693 Chr.13: 31246977-31246977 on Build GRCh38
rs142421716 Chr.13: 31247096-31247096 on Build GRCh38
rs967997643 Chr.13: 31246955-31246955 on Build GRCh38
rs774732536 Chr.13: 31247137-31247137 on Build GRCh38
rs199535028 Chr.13: 31246955-31246956 on Build GRCh38
rs947477122 Chr.13: 31246760-31246760 on Build GRCh38
rs1026053238 Chr.13: 31246835-31246835 on Build GRCh38
rs375070601 Chr.13: 31246967-31246968 on Build GRCh38
rs749572751 Chr.13: 31247020-31247020 on Build GRCh38
rs79127728 Chr.13: 31246985-31246985 on Build GRCh38
rs751146291 Chr.13: 31247088-31247088 on Build GRCh38
rs367941054 Chr.13: 31246995-31246995 on Build GRCh38
rs766371402 Chr.13: 31246973-31246973 on Build GRCh38
rs9542300 Chr.13: 31246956-31246956 on Build GRCh38
rs117111131 Chr.13: 31247011-31247011 on Build GRCh38
rs761989807 Chr.13: 31247041-31247041 on Build GRCh38
rs1056372825 Chr.13: 31246728-31246728 on Build GRCh38
rs369000249 Chr.13: 31247109-31247109 on Build GRCh38
rs1038759277 Chr.13: 31246812-31246812 on Build GRCh38
rs549964753 Chr.13: 31246801-31246801 on Build GRCh38
rs765118210 Chr.13: 31247080-31247080 on Build GRCh38
rs781089564 Chr.13: 31247026-31247026 on Build GRCh38
rs780562438 Chr.13: 31247114-31247114 on Build GRCh38
rs202065963 Chr.13: 31246998-31246998 on Build GRCh38
rs116187973 Chr.13: 31246881-31246881 on Build GRCh38
rs748371730 Chr.13: 31247139-31247139 on Build GRCh38
rs186499393 Chr.13: 31246937-31246937 on Build GRCh38
rs1924822 Chr.13: 31246792-31246792 on Build GRCh38
rs9542307 Chr.13: 31247119-31247119 on Build GRCh38
rs773136617 Chr.13: 31247147-31247147 on Build GRCh38
rs66466340 Chr.13: 31246951-31246952 on Build GRCh38
rs772263738 Chr.13: 31247142-31247142 on Build GRCh38
rs9592712 Chr.13: 31246929-31246929 on Build GRCh38
rs890380993 Chr.13: 31246799-31246799 on Build GRCh38
rs777311145 Chr.13: 31246987-31246987 on Build GRCh38
rs1002142694 Chr.13: 31247144-31247144 on Build GRCh38
rs368196937 Chr.13: 31246941-31246941 on Build GRCh38
rs183322816 Chr.13: 31247015-31247015 on Build GRCh38
rs761936753 Chr.13: 31246972-31246972 on Build GRCh38
rs9564692 Chr.13: 31247103-31247103 on Build GRCh38
rs545131155 Chr.13: 31246934-31246934 on Build GRCh38
rs1002490452 Chr.13: 31246961-31246961 on Build GRCh38
rs759565170 Chr.13: 31247149-31247149 on Build GRCh38
rs908558949 Chr.13: 31246793-31246793 on Build GRCh38
rs531073276 Chr.13: 31246946-31246946 on Build GRCh38
rs200933165 Chr.13: 31246954-31246955 on Build GRCh38
rs1034044148 Chr.13: 31247152-31247152 on Build GRCh38
rs561301280 Chr.13: 31246976-31246976 on Build GRCh38
rs758136161 Chr.13: 31247086-31247086 on Build GRCh38
rs1017875276 Chr.13: 31246804-31246804 on Build GRCh38
rs752463879 Chr.13: 31247084-31247084 on Build GRCh38
rs916077071 Chr.13: 31246932-31246936 on Build GRCh38
rs9542305 Chr.13: 31247040-31247040 on Build GRCh38
rs769023873 Chr.13: 31247024-31247024 on Build GRCh38
rs34698943 Chr.13: 31247014-31247014 on Build GRCh38
rs577783560 Chr.13: 31246932-31246932 on Build GRCh38
rs927577920 Chr.13: 31246927-31246929 on Build GRCh38
rs749702385 Chr.13: 31247116-31247116 on Build GRCh38
rs926414152 Chr.13: 31246753-31246753 on Build GRCh38
rs199976217 Chr.13: 31246946-31246947 on Build GRCh38
rs924691522 Chr.13: 31246944-31246951 on Build GRCh38
rs991731861 Chr.13: 31246929-31246930 on Build GRCh38
rs77790387 Chr.13: 31246936-31246936 on Build GRCh38
rs186315516 Chr.13: 31246726-31246726 on Build GRCh38
rs1008613665 Chr.13: 31246892-31246892 on Build GRCh38
rs903220468 Chr.13: 31246771-31246771 on Build GRCh38
rs181469485 Chr.13: 31246883-31246883 on Build GRCh38
rs747050938 Chr.13: 31247028-31247028 on Build GRCh38
rs191897907 Chr.13: 31246764-31246764 on Build GRCh38
rs780553393 Chr.13: 31247097-31247097 on Build GRCh38
rs1043146165 Chr.13: 31246939-31246939 on Build GRCh38
rs535077829 Chr.13: 31246803-31246803 on Build GRCh38
rs980662548 Chr.13: 31246734-31246734 on Build GRCh38
rs137993898 Chr.13: 31247072-31247072 on Build GRCh38
rs368693579 Chr.13: 31247049-31247049 on Build GRCh38
rs746302670 Chr.13: 31247002-31247002 on Build GRCh38
rs932889748 Chr.13: 31246828-31246828 on Build GRCh38
rs746541374 Chr.13: 31246991-31246991 on Build GRCh38
rs374596352 Chr.13: 31246967-31246967 on Build GRCh38
rs1034702493 Chr.13: 31247143-31247143 on Build GRCh38

View Full Product Details