Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 40807208-40807707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A15
Entrez Gene ID
10166
Gene Name
solute carrier family 25 member 15
Gene Aliases
D13S327, HHH, ORC1, ORNT1
UniGene
Hs.646645
Gene Chromosome Location
Chr.13: 40789411-40812460 on Build GRCh38
Transcripts
NM_014252.3 NR_038258.1 NR_038259.1

Gene Symbol
TPTE2P5
Entrez Gene ID
100616668
Gene Name
transmembrane phosphoinositide 3-phosphatase and tensin homolog 2 pseudogene 5
Gene Aliases
-
UniGene
Hs.660718
Gene Chromosome Location
Chr.13: 40796985-40921750 on Build GRCh38
Transcripts
NM_014252.3 NR_038258.1 NR_038259.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3417589 c.536G>A p.R179Q Chr.13: 40807377-40807377 on Build GRCh38
469425 c.614A>G p.D205G Chr.13: 40807455-40807455 on Build GRCh38
3724776 c.492T>C p.D164D Chr.13: 40807333-40807333 on Build GRCh38
6259594 c.569G>T p.G190V Chr.13: 40807410-40807410 on Build GRCh38
6448343 c.583C>T p.R195W Chr.13: 40807424-40807424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141028076 Chr.13: 40807405-40807405 on Build GRCh38
rs1022470320 Chr.13: 40807542-40807542 on Build GRCh38
rs538507004 Chr.13: 40807267-40807267 on Build GRCh38
rs762025172 Chr.13: 40807471-40807471 on Build GRCh38
rs745823100 Chr.13: 40807284-40807284 on Build GRCh38
rs762064681 Chr.13: 40807510-40807510 on Build GRCh38
rs770355365 Chr.13: 40807425-40807425 on Build GRCh38
rs200117115 Chr.13: 40807299-40807299 on Build GRCh38
rs769552929 Chr.13: 40807508-40807509 on Build GRCh38
rs576852849 Chr.13: 40807458-40807458 on Build GRCh38
rs760502972 Chr.13: 40807494-40807494 on Build GRCh38
rs759984675 Chr.13: 40807344-40807344 on Build GRCh38
rs202247803 Chr.13: 40807403-40807405 on Build GRCh38
rs778022395 Chr.13: 40807514-40807514 on Build GRCh38
rs545623290 Chr.13: 40807554-40807554 on Build GRCh38
rs767127481 Chr.13: 40807473-40807473 on Build GRCh38
rs756007568 Chr.13: 40807389-40807389 on Build GRCh38
rs1020343088 Chr.13: 40807560-40807560 on Build GRCh38
rs375061803 Chr.13: 40807272-40807272 on Build GRCh38
rs927467691 Chr.13: 40807233-40807233 on Build GRCh38
rs769371469 Chr.13: 40807319-40807319 on Build GRCh38
rs104894424 Chr.13: 40807379-40807379 on Build GRCh38
rs772487311 Chr.13: 40807307-40807307 on Build GRCh38
rs531613734 Chr.13: 40807275-40807275 on Build GRCh38
rs1013910201 Chr.13: 40807516-40807516 on Build GRCh38
rs978299443 Chr.13: 40807237-40807237 on Build GRCh38
rs111306278 Chr.13: 40807569-40807569 on Build GRCh38
rs758060621 Chr.13: 40807273-40807273 on Build GRCh38
rs989205841 Chr.13: 40807541-40807541 on Build GRCh38
rs748169316 Chr.13: 40807289-40807289 on Build GRCh38
rs772977811 Chr.13: 40807643-40807643 on Build GRCh38
rs200321784 Chr.13: 40807354-40807354 on Build GRCh38
rs1048233546 Chr.13: 40807363-40807363 on Build GRCh38
rs796052041 Chr.13: 40807347-40807347 on Build GRCh38
rs373477060 Chr.13: 40807515-40807515 on Build GRCh38
rs771173592 Chr.13: 40807330-40807330 on Build GRCh38
rs911583253 Chr.13: 40807654-40807654 on Build GRCh38
rs777014493 Chr.13: 40807331-40807331 on Build GRCh38
rs368094314 Chr.13: 40807286-40807286 on Build GRCh38
rs374352017 Chr.13: 40807393-40807393 on Build GRCh38
rs778131013 Chr.13: 40807323-40807323 on Build GRCh38
rs747458822 Chr.13: 40807328-40807328 on Build GRCh38
rs978303386 Chr.13: 40807637-40807637 on Build GRCh38
rs766476051 Chr.13: 40807498-40807498 on Build GRCh38
rs1038825312 Chr.13: 40807327-40807327 on Build GRCh38
rs754930419 Chr.13: 40807509-40807509 on Build GRCh38
rs1001813690 Chr.13: 40807438-40807438 on Build GRCh38
rs753799938 Chr.13: 40807507-40807507 on Build GRCh38
rs957788324 Chr.13: 40807464-40807464 on Build GRCh38
rs202247804 Chr.13: 40807410-40807410 on Build GRCh38
rs151239794 Chr.13: 40807406-40807406 on Build GRCh38
rs370639849 Chr.13: 40807266-40807266 on Build GRCh38
rs117385445 Chr.13: 40807665-40807665 on Build GRCh38
rs781425166 Chr.13: 40807421-40807421 on Build GRCh38
rs773862892 Chr.13: 40807457-40807457 on Build GRCh38
rs967007588 Chr.13: 40807584-40807585 on Build GRCh38
rs751412119 Chr.13: 40807274-40807274 on Build GRCh38
rs746179010 Chr.13: 40807422-40807422 on Build GRCh38
rs774986959 Chr.13: 40807348-40807348 on Build GRCh38
rs761119245 Chr.13: 40807263-40807263 on Build GRCh38
rs911800976 Chr.13: 40807297-40807297 on Build GRCh38
rs770229041 Chr.13: 40807345-40807345 on Build GRCh38
rs774514708 Chr.13: 40807417-40807417 on Build GRCh38
rs372332143 Chr.13: 40807303-40807303 on Build GRCh38
rs768185714 Chr.13: 40807449-40807449 on Build GRCh38
rs966518982 Chr.13: 40807556-40807556 on Build GRCh38
rs767241202 Chr.13: 40807563-40807563 on Build GRCh38
rs775952093 Chr.13: 40807426-40807426 on Build GRCh38
rs200873328 Chr.13: 40807362-40807362 on Build GRCh38
rs762489495 Chr.13: 40807442-40807442 on Build GRCh38
rs959137773 Chr.13: 40807253-40807253 on Build GRCh38
rs750249238 Chr.13: 40807493-40807493 on Build GRCh38
rs104894429 Chr.13: 40807376-40807376 on Build GRCh38
rs781229082 Chr.13: 40807278-40807278 on Build GRCh38
rs770194398 Chr.13: 40807265-40807265 on Build GRCh38
rs747230863 Chr.13: 40807409-40807409 on Build GRCh38
rs750171185 Chr.13: 40807382-40807382 on Build GRCh38

View Full Product Details