Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 50943129-50943625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNASEH2B
Entrez Gene ID
79621
Gene Name
ribonuclease H2 subunit B
Gene Aliases
AGS2, DLEU8
UniGene
Hs.306291
Gene Chromosome Location
Chr.13: 50909678-50970462 on Build GRCh38
Transcripts
NM_001142279.2 NM_024570.3 XM_006719867.3 XM_011535229.1 XM_011535230.2 XM_011535231.2 XM_011535233.2 XM_017020747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1639366 c.503delA p.K170fs*9 Chr.13: 50943387-50943387 on Build GRCh38
7308184 c.503A>G p.E168G Chr.13: 50943387-50943387 on Build GRCh38
1942875 c.459G>T p.K153N Chr.13: 50943343-50943343 on Build GRCh38
6767100 c.498G>A p.W166* Chr.13: 50943382-50943382 on Build GRCh38
6045901 c.467A>G p.Y156C Chr.13: 50943351-50943351 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777313709 Chr.13: 50943320-50943320 on Build GRCh38
rs760216248 Chr.13: 50943409-50943410 on Build GRCh38
rs374276957 Chr.13: 50943292-50943292 on Build GRCh38
rs778136566 Chr.13: 50943445-50943445 on Build GRCh38
rs989812532 Chr.13: 50943449-50943449 on Build GRCh38
rs1003145341 Chr.13: 50943277-50943277 on Build GRCh38
rs775381857 Chr.13: 50943304-50943305 on Build GRCh38
rs908748638 Chr.13: 50943563-50943563 on Build GRCh38
rs577412841 Chr.13: 50943212-50943212 on Build GRCh38
rs1011904226 Chr.13: 50943354-50943354 on Build GRCh38
rs980849182 Chr.13: 50943492-50943492 on Build GRCh38
rs75971463 Chr.13: 50943369-50943369 on Build GRCh38
rs753208462 Chr.13: 50943428-50943428 on Build GRCh38
rs528697643 Chr.13: 50943286-50943286 on Build GRCh38
rs559675078 Chr.13: 50943266-50943266 on Build GRCh38
rs1047246863 Chr.13: 50943255-50943255 on Build GRCh38
rs769721789 Chr.13: 50943342-50943342 on Build GRCh38
rs989495791 Chr.13: 50943153-50943153 on Build GRCh38
rs775300215 Chr.13: 50943352-50943352 on Build GRCh38
rs1035038442 Chr.13: 50943294-50943294 on Build GRCh38
rs780141262 Chr.13: 50943283-50943283 on Build GRCh38
rs762711560 Chr.13: 50943367-50943367 on Build GRCh38
rs576591610 Chr.13: 50943169-50943169 on Build GRCh38
rs545069822 Chr.13: 50943277-50943277 on Build GRCh38
rs372499426 Chr.13: 50943433-50943433 on Build GRCh38
rs771902207 Chr.13: 50943313-50943313 on Build GRCh38
rs746633003 Chr.13: 50943324-50943324 on Build GRCh38
rs1030880550 Chr.13: 50943401-50943401 on Build GRCh38
rs146451037 Chr.13: 50943339-50943339 on Build GRCh38
rs753938285 Chr.13: 50943405-50943405 on Build GRCh38
rs1010696077 Chr.13: 50943217-50943217 on Build GRCh38
rs960799762 Chr.13: 50943509-50943509 on Build GRCh38
rs763896138 Chr.13: 50943382-50943382 on Build GRCh38
rs928110769 Chr.13: 50943242-50943242 on Build GRCh38
rs563712972 Chr.13: 50943185-50943185 on Build GRCh38
rs941586397 Chr.13: 50943596-50943596 on Build GRCh38
rs983575972 Chr.13: 50943418-50943418 on Build GRCh38
rs981267730 Chr.13: 50943230-50943230 on Build GRCh38
rs1028760869 Chr.13: 50943312-50943312 on Build GRCh38
rs760884054 Chr.13: 50943390-50943390 on Build GRCh38
rs546345797 Chr.13: 50943243-50943243 on Build GRCh38
rs138213208 Chr.13: 50943471-50943471 on Build GRCh38
rs79310911 Chr.13: 50943372-50943372 on Build GRCh38
rs966293199 Chr.13: 50943458-50943458 on Build GRCh38
rs202232001 Chr.13: 50943349-50943349 on Build GRCh38
rs745657062 Chr.13: 50943276-50943277 on Build GRCh38
rs549365464 Chr.13: 50943546-50943546 on Build GRCh38
rs770355902 Chr.13: 50943329-50943329 on Build GRCh38
rs898833386 Chr.13: 50943298-50943298 on Build GRCh38
rs78315883 Chr.13: 50943387-50943387 on Build GRCh38
rs77301371 Chr.13: 50943395-50943395 on Build GRCh38
rs1927742 Chr.13: 50943463-50943463 on Build GRCh38
rs756167483 Chr.13: 50943271-50943271 on Build GRCh38
rs748494404 Chr.13: 50943311-50943311 on Build GRCh38
rs928036967 Chr.13: 50943508-50943508 on Build GRCh38
rs1054451672 Chr.13: 50943159-50943159 on Build GRCh38
rs765819204 Chr.13: 50943412-50943412 on Build GRCh38
rs373236020 Chr.13: 50943207-50943207 on Build GRCh38
rs759529625 Chr.13: 50943340-50943340 on Build GRCh38
rs772054107 Chr.13: 50943305-50943305 on Build GRCh38
rs766382476 Chr.13: 50943396-50943396 on Build GRCh38
rs755608455 Chr.13: 50943406-50943406 on Build GRCh38
rs755493007 Chr.13: 50943274-50943275 on Build GRCh38
rs76219783 Chr.13: 50943360-50943360 on Build GRCh38
rs779476919 Chr.13: 50943301-50943301 on Build GRCh38

View Full Product Details