Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 50949385-50949887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNASEH2B
Entrez Gene ID
79621
Gene Name
ribonuclease H2 subunit B
Gene Aliases
AGS2, DLEU8
UniGene
Hs.306291
Gene Chromosome Location
Chr.13: 50909678-50970462 on Build GRCh38
Transcripts
NM_001142279.2 NM_024570.3 XM_006719867.3 XM_011535229.1 XM_011535230.2 XM_011535231.2 XM_011535233.2 XM_017020747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6201477 c.705_707delAGA p.E236delE Chr.13: 50949469-50949471 on Build GRCh38
389103 c.699G>T p.K233N Chr.13: 50949463-50949463 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768783637 Chr.13: 50949463-50949463 on Build GRCh38
rs751497437 Chr.13: 50949547-50949547 on Build GRCh38
rs957957502 Chr.13: 50949717-50949717 on Build GRCh38
rs924104958 Chr.13: 50949705-50949705 on Build GRCh38
rs753266057 Chr.13: 50949520-50949520 on Build GRCh38
rs945220894 Chr.13: 50949464-50949464 on Build GRCh38
rs1057269037 Chr.13: 50949843-50949843 on Build GRCh38
rs963332273 Chr.13: 50949528-50949528 on Build GRCh38
rs981945320 Chr.13: 50949763-50949763 on Build GRCh38
rs764105023 Chr.13: 50949542-50949542 on Build GRCh38
rs774234566 Chr.13: 50949469-50949469 on Build GRCh38
rs139886655 Chr.13: 50949685-50949685 on Build GRCh38
rs746773572 Chr.13: 50949430-50949430 on Build GRCh38
rs749427005 Chr.13: 50949443-50949443 on Build GRCh38
rs571254594 Chr.13: 50949559-50949559 on Build GRCh38
rs751061303 Chr.13: 50949580-50949580 on Build GRCh38
rs994824023 Chr.13: 50949593-50949593 on Build GRCh38
rs937967322 Chr.13: 50949789-50949789 on Build GRCh38
rs200081827 Chr.13: 50949536-50949536 on Build GRCh38
rs1046177737 Chr.13: 50949485-50949485 on Build GRCh38
rs964991022 Chr.13: 50949686-50949686 on Build GRCh38
rs1009071314 Chr.13: 50949672-50949672 on Build GRCh38
rs913657824 Chr.13: 50949736-50949736 on Build GRCh38
rs187099374 Chr.13: 50949413-50949413 on Build GRCh38
rs754489489 Chr.13: 50949762-50949762 on Build GRCh38
rs190327832 Chr.13: 50949498-50949498 on Build GRCh38
rs948314085 Chr.13: 50949744-50949744 on Build GRCh38
rs748746326 Chr.13: 50949471-50949471 on Build GRCh38
rs776350813 Chr.13: 50949533-50949533 on Build GRCh38
rs747656006 Chr.13: 50949761-50949762 on Build GRCh38
rs773891895 Chr.13: 50949759-50949759 on Build GRCh38
rs770193650 Chr.13: 50949434-50949434 on Build GRCh38
rs796344476 Chr.13: 50949664-50949664 on Build GRCh38
rs752694584 Chr.13: 50949421-50949421 on Build GRCh38
rs950619022 Chr.13: 50949431-50949431 on Build GRCh38
rs1028009844 Chr.13: 50949711-50949711 on Build GRCh38
rs758398563 Chr.13: 50949422-50949422 on Build GRCh38
rs879095085 Chr.13: 50949556-50949556 on Build GRCh38
rs764862165 Chr.13: 50949534-50949534 on Build GRCh38
rs1048289570 Chr.13: 50949649-50949649 on Build GRCh38
rs996816428 Chr.13: 50949710-50949710 on Build GRCh38
rs745710166 Chr.13: 50949536-50949553 on Build GRCh38
rs780442662 Chr.13: 50949441-50949441 on Build GRCh38
rs533617308 Chr.13: 50949575-50949575 on Build GRCh38
rs570354333 Chr.13: 50949415-50949415 on Build GRCh38
rs777780324 Chr.13: 50949429-50949429 on Build GRCh38
rs1055161129 Chr.13: 50949566-50949566 on Build GRCh38
rs375015766 Chr.13: 50949525-50949525 on Build GRCh38
rs989128216 Chr.13: 50949723-50949723 on Build GRCh38
rs886488578 Chr.13: 50949658-50949658 on Build GRCh38
rs1053919140 Chr.13: 50949726-50949726 on Build GRCh38
rs573451915 Chr.13: 50949718-50949718 on Build GRCh38
rs568641923 Chr.13: 50949767-50949767 on Build GRCh38
rs372632599 Chr.13: 50949483-50949483 on Build GRCh38
rs761005167 Chr.13: 50949484-50949484 on Build GRCh38
rs1002595940 Chr.13: 50949795-50949795 on Build GRCh38
rs886050290 Chr.13: 50949452-50949452 on Build GRCh38
rs954779362 Chr.13: 50949656-50949656 on Build GRCh38
rs927925747 Chr.13: 50949532-50949532 on Build GRCh38
rs780451460 Chr.13: 50949564-50949564 on Build GRCh38
rs886215243 Chr.13: 50949855-50949855 on Build GRCh38
rs574227210 Chr.13: 50949797-50949797 on Build GRCh38
rs369970613 Chr.13: 50949417-50949417 on Build GRCh38

View Full Product Details