Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 50953767-50954263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNASEH2B
Entrez Gene ID
79621
Gene Name
ribonuclease H2 subunit B
Gene Aliases
AGS2, DLEU8
UniGene
Hs.306291
Gene Chromosome Location
Chr.13: 50909678-50970462 on Build GRCh38
Transcripts
NM_001142279.2 NM_024570.3 XM_006719867.3 XM_011535229.1 XM_011535233.2 XM_017020747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6404930 c.761A>G p.E254G Chr.13: 50953924-50953924 on Build GRCh38
7325110 c.770A>G p.E257G Chr.13: 50953933-50953933 on Build GRCh38
1300243 c.787A>G p.T263A Chr.13: 50953950-50953950 on Build GRCh38
328852 c.777A>T p.K259N Chr.13: 50953940-50953940 on Build GRCh38
6139073 c.789T>C p.T263T Chr.13: 50953952-50953952 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761333168 Chr.13: 50953990-50953990 on Build GRCh38
rs139840761 Chr.13: 50954153-50954153 on Build GRCh38
rs189116235 Chr.13: 50954029-50954029 on Build GRCh38
rs775119865 Chr.13: 50953971-50953971 on Build GRCh38
rs749536701 Chr.13: 50953868-50953868 on Build GRCh38
rs866929388 Chr.13: 50953847-50953847 on Build GRCh38
rs755138594 Chr.13: 50953885-50953885 on Build GRCh38
rs369199159 Chr.13: 50954015-50954015 on Build GRCh38
rs187170706 Chr.13: 50954155-50954155 on Build GRCh38
rs993105829 Chr.13: 50953796-50953796 on Build GRCh38
rs778980232 Chr.13: 50954014-50954014 on Build GRCh38
rs750165625 Chr.13: 50954160-50954160 on Build GRCh38
rs981814910 Chr.13: 50954121-50954127 on Build GRCh38
rs767399621 Chr.13: 50954000-50954000 on Build GRCh38
rs747575443 Chr.13: 50954023-50954023 on Build GRCh38
rs779113809 Chr.13: 50953893-50953893 on Build GRCh38
rs1013988153 Chr.13: 50954147-50954147 on Build GRCh38
rs780138105 Chr.13: 50954206-50954206 on Build GRCh38
rs777772670 Chr.13: 50954022-50954022 on Build GRCh38
rs183734174 Chr.13: 50953951-50953951 on Build GRCh38
rs776847998 Chr.13: 50953949-50953949 on Build GRCh38
rs763654606 Chr.13: 50953981-50953981 on Build GRCh38
rs183787536 Chr.13: 50954137-50954137 on Build GRCh38
rs750179605 Chr.13: 50954001-50954001 on Build GRCh38
rs998133701 Chr.13: 50954208-50954209 on Build GRCh38
rs999858195 Chr.13: 50954079-50954079 on Build GRCh38
rs76413207 Chr.13: 50953991-50953991 on Build GRCh38
rs371349565 Chr.13: 50953867-50953867 on Build GRCh38
rs891576449 Chr.13: 50954142-50954142 on Build GRCh38
rs969929424 Chr.13: 50954159-50954159 on Build GRCh38
rs1052712728 Chr.13: 50954107-50954107 on Build GRCh38
rs576231348 Chr.13: 50954171-50954173 on Build GRCh38
rs373179837 Chr.13: 50954240-50954240 on Build GRCh38
rs146378238 Chr.13: 50953925-50953925 on Build GRCh38
rs756850979 Chr.13: 50953887-50953887 on Build GRCh38
rs771299667 Chr.13: 50953929-50953929 on Build GRCh38
rs530808995 Chr.13: 50954169-50954169 on Build GRCh38
rs748144224 Chr.13: 50953907-50953907 on Build GRCh38
rs970403543 Chr.13: 50954097-50954097 on Build GRCh38
rs757200421 Chr.13: 50953993-50953993 on Build GRCh38
rs1024129566 Chr.13: 50953825-50953825 on Build GRCh38
rs150363383 Chr.13: 50953950-50953950 on Build GRCh38
rs1048219423 Chr.13: 50954161-50954161 on Build GRCh38
rs917519640 Chr.13: 50954064-50954064 on Build GRCh38
rs755266257 Chr.13: 50954007-50954007 on Build GRCh38
rs1018669360 Chr.13: 50953814-50953814 on Build GRCh38
rs962615193 Chr.13: 50954174-50954174 on Build GRCh38
rs1043966687 Chr.13: 50953999-50953999 on Build GRCh38
rs1006811280 Chr.13: 50953810-50953810 on Build GRCh38
rs972687891 Chr.13: 50953936-50953936 on Build GRCh38
rs146607498 Chr.13: 50954184-50954184 on Build GRCh38
rs780778643 Chr.13: 50953865-50953865 on Build GRCh38
rs1039604712 Chr.13: 50954195-50954195 on Build GRCh38
rs747450548 Chr.13: 50953926-50953926 on Build GRCh38
rs757145306 Chr.13: 50953858-50953858 on Build GRCh38
rs191265144 Chr.13: 50954030-50954030 on Build GRCh38
rs113827426 Chr.13: 50954035-50954035 on Build GRCh38
rs113879083 Chr.13: 50953835-50953836 on Build GRCh38
rs200721645 Chr.13: 50953919-50953919 on Build GRCh38
rs942585012 Chr.13: 50954188-50954188 on Build GRCh38

View Full Product Details