Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21310459-21310965 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPGRIP1
Entrez Gene ID
57096
Gene Name
retinitis pigmentosa GTPase regulator interacting protein 1
Gene Aliases
CORD13, LCA6, RGI1, RGRIP, RPGRIP, RPGRIP1d
UniGene
Hs.126035
Gene Chromosome Location
Chr.14: 21287977-21351316 on Build GRCh38
Transcripts
NM_020366.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6404158 c.907G>T p.A303S Chr.14: 21310584-21310584 on Build GRCh38
5382050 c.918C>T p.T306T Chr.14: 21310595-21310595 on Build GRCh38
5432441 c.912C>T p.Y304Y Chr.14: 21310589-21310589 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs964840457 Chr.14: 21310788-21310788 on Build GRCh38
rs181663715 Chr.14: 21310801-21310801 on Build GRCh38
rs373627836 Chr.14: 21310619-21310619 on Build GRCh38
rs1030689150 Chr.14: 21310861-21310861 on Build GRCh38
rs747880709 Chr.14: 21310632-21310632 on Build GRCh38
rs774179253 Chr.14: 21310648-21310648 on Build GRCh38
rs1019211103 Chr.14: 21310764-21310764 on Build GRCh38
rs370221426 Chr.14: 21310671-21310671 on Build GRCh38
rs150107283 Chr.14: 21310610-21310610 on Build GRCh38
rs960556537 Chr.14: 21310867-21310867 on Build GRCh38
rs772595382 Chr.14: 21310555-21310555 on Build GRCh38
rs780626446 Chr.14: 21310928-21310928 on Build GRCh38
rs777964919 Chr.14: 21310596-21310596 on Build GRCh38
rs759228419 Chr.14: 21310794-21310794 on Build GRCh38
rs751011542 Chr.14: 21310922-21310922 on Build GRCh38
rs940651328 Chr.14: 21310936-21310936 on Build GRCh38
rs756323674 Chr.14: 21310591-21310591 on Build GRCh38
rs568675254 Chr.14: 21310573-21310573 on Build GRCh38
rs960505008 Chr.14: 21310533-21310533 on Build GRCh38
rs766243294 Chr.14: 21310582-21310582 on Build GRCh38
rs542161968 Chr.14: 21310933-21310933 on Build GRCh38
rs762375739 Chr.14: 21310639-21310639 on Build GRCh38
rs983846930 Chr.14: 21310863-21310863 on Build GRCh38
rs779025532 Chr.14: 21310914-21310914 on Build GRCh38
rs1012201448 Chr.14: 21310783-21310783 on Build GRCh38
rs138519972 Chr.14: 21310935-21310935 on Build GRCh38
rs200168103 Chr.14: 21310584-21310584 on Build GRCh38
rs201852827 Chr.14: 21310900-21310900 on Build GRCh38
rs777323892 Chr.14: 21310939-21310939 on Build GRCh38
rs766807499 Chr.14: 21310826-21310826 on Build GRCh38
rs879573531 Chr.14: 21310766-21310766 on Build GRCh38
rs1957414 Chr.14: 21310633-21310633 on Build GRCh38
rs752474118 Chr.14: 21310866-21310866 on Build GRCh38
rs200700552 Chr.14: 21310644-21310644 on Build GRCh38
rs1000250149 Chr.14: 21310658-21310658 on Build GRCh38
rs746332573 Chr.14: 21310617-21310617 on Build GRCh38
rs757291015 Chr.14: 21310603-21310603 on Build GRCh38
rs398099213 Chr.14: 21310568-21310570 on Build GRCh38
rs1047742693 Chr.14: 21310537-21310537 on Build GRCh38
rs202088735 Chr.14: 21310823-21310823 on Build GRCh38
rs568372341 Chr.14: 21310589-21310589 on Build GRCh38
rs991996158 Chr.14: 21310876-21310876 on Build GRCh38
rs999249944 Chr.14: 21310829-21310829 on Build GRCh38
rs368531749 Chr.14: 21310941-21310941 on Build GRCh38
rs377350887 Chr.14: 21310579-21310579 on Build GRCh38
rs780538730 Chr.14: 21310630-21310630 on Build GRCh38
rs759437512 Chr.14: 21310649-21310649 on Build GRCh38
rs373013143 Chr.14: 21310551-21310551 on Build GRCh38
rs752546353 Chr.14: 21310915-21310915 on Build GRCh38
rs765030557 Chr.14: 21310860-21310860 on Build GRCh38
rs772866174 Chr.14: 21310638-21310638 on Build GRCh38
rs556812237 Chr.14: 21310872-21310872 on Build GRCh38
rs746182338 Chr.14: 21310627-21310628 on Build GRCh38
rs757285269 Chr.14: 21310686-21310690 on Build GRCh38
rs375024142 Chr.14: 21310697-21310697 on Build GRCh38
rs956480934 Chr.14: 21310695-21310695 on Build GRCh38
rs754799432 Chr.14: 21310585-21310585 on Build GRCh38
rs758973246 Chr.14: 21310924-21310924 on Build GRCh38
rs375834040 Chr.14: 21310660-21310660 on Build GRCh38
rs752956324 Chr.14: 21310590-21310590 on Build GRCh38
rs764224388 Chr.14: 21310897-21310897 on Build GRCh38
rs772641361 Chr.14: 21310895-21310896 on Build GRCh38
rs201864245 Chr.14: 21310767-21310767 on Build GRCh38
rs939984565 Chr.14: 21310562-21310562 on Build GRCh38
rs749340109 Chr.14: 21310597-21310597 on Build GRCh38
rs767365924 Chr.14: 21310865-21310865 on Build GRCh38
rs377504054 Chr.14: 21310747-21310747 on Build GRCh38
rs757303340 Chr.14: 21310908-21310908 on Build GRCh38

View Full Product Details