Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000633.2 | NP_000624.2 | 2 - 3 | 1081 | 96 |
GenBank mRNA | BC027258.1 | - | 2 - 3 | 977 | 96 |
EU287875.1 | - | 1 - 2 | 588 | 96 | |
M13994.1 | - | 1 - 2 | 2046 | 96 | |
M14745.1 | - | 1 - 2 | 619 | 96 | |
X06487.1 | - | 2 - 3 | 1474 | 96 |