Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 23835851-23836350 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRKCB
Entrez Gene ID
5579
Gene Name
protein kinase C beta
Gene Aliases
PKC-beta, PKCB, PRKCB1, PRKCB2
UniGene
Hs.460355
Gene Chromosome Location
Chr.16: 23835979-24220611 on Build GRCh38
Transcripts
NM_002738.6 NM_212535.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5706049 c.64C>G p.R22G Chr.16: 23836239-23836239 on Build GRCh38
3728246 c.149G>C p.C50S Chr.16: 23836324-23836324 on Build GRCh38
3280072 c.102C>G p.V34V Chr.16: 23836277-23836277 on Build GRCh38
6755778 c.93G>A p.V31V Chr.16: 23836268-23836268 on Build GRCh38
3280063 c.51C>A p.T17T Chr.16: 23836226-23836226 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974336719 Chr.16: 23835874-23835874 on Build GRCh38
rs1054309339 Chr.16: 23835937-23835937 on Build GRCh38
rs996053363 Chr.16: 23835986-23835986 on Build GRCh38
rs773819749 Chr.16: 23836225-23836225 on Build GRCh38
rs896914892 Chr.16: 23835977-23835977 on Build GRCh38
rs990351629 Chr.16: 23836038-23836038 on Build GRCh38
rs890774772 Chr.16: 23836121-23836121 on Build GRCh38
rs959665377 Chr.16: 23835888-23835889 on Build GRCh38
rs370984906 Chr.16: 23836210-23836210 on Build GRCh38
rs767070226 Chr.16: 23836152-23836152 on Build GRCh38
rs943676785 Chr.16: 23836125-23836125 on Build GRCh38
rs75317242 Chr.16: 23835904-23835904 on Build GRCh38
rs553428223 Chr.16: 23835944-23835944 on Build GRCh38
rs996778422 Chr.16: 23835942-23835942 on Build GRCh38
rs759234204 Chr.16: 23836231-23836231 on Build GRCh38
rs757085223 Chr.16: 23836186-23836186 on Build GRCh38
rs201366670 Chr.16: 23836189-23836189 on Build GRCh38
rs753829465 Chr.16: 23836253-23836253 on Build GRCh38
rs1042535127 Chr.16: 23835920-23835920 on Build GRCh38
rs756077698 Chr.16: 23836163-23836163 on Build GRCh38
rs751296237 Chr.16: 23836144-23836144 on Build GRCh38
rs1036672437 Chr.16: 23836138-23836138 on Build GRCh38
rs893020697 Chr.16: 23835939-23835939 on Build GRCh38
rs75964872 Chr.16: 23836254-23836254 on Build GRCh38
rs775089575 Chr.16: 23836241-23836241 on Build GRCh38
rs1015321832 Chr.16: 23835975-23835975 on Build GRCh38
rs868538925 Chr.16: 23835988-23835988 on Build GRCh38
rs575795787 Chr.16: 23836200-23836200 on Build GRCh38
rs1049673781 Chr.16: 23836114-23836114 on Build GRCh38
rs751689932 Chr.16: 23836274-23836274 on Build GRCh38
rs1024977054 Chr.16: 23835990-23835990 on Build GRCh38
rs535808203 Chr.16: 23836156-23836156 on Build GRCh38
rs776684827 Chr.16: 23835913-23835913 on Build GRCh38
rs766899029 Chr.16: 23836233-23836233 on Build GRCh38
rs932555926 Chr.16: 23836087-23836087 on Build GRCh38
rs368093414 Chr.16: 23836201-23836201 on Build GRCh38
rs924419933 Chr.16: 23835930-23835930 on Build GRCh38
rs763455165 Chr.16: 23836142-23836142 on Build GRCh38
rs146177740 Chr.16: 23836262-23836262 on Build GRCh38
rs749669754 Chr.16: 23836213-23836213 on Build GRCh38
rs923993890 Chr.16: 23836022-23836022 on Build GRCh38
rs760228327 Chr.16: 23836245-23836245 on Build GRCh38
rs777496943 Chr.16: 23836166-23836166 on Build GRCh38
rs188342168 Chr.16: 23836151-23836151 on Build GRCh38
rs781315165 Chr.16: 23836304-23836304 on Build GRCh38
rs1012485614 Chr.16: 23836185-23836185 on Build GRCh38
rs768549392 Chr.16: 23836193-23836193 on Build GRCh38
rs890828940 Chr.16: 23835949-23835949 on Build GRCh38
rs1031003748 Chr.16: 23836313-23836313 on Build GRCh38
rs781362818 Chr.16: 23836198-23836198 on Build GRCh38
rs763989321 Chr.16: 23836247-23836247 on Build GRCh38
rs771388864 Chr.16: 23836215-23836215 on Build GRCh38
rs914421554 Chr.16: 23836061-23836061 on Build GRCh38
rs1007931397 Chr.16: 23835962-23835962 on Build GRCh38
rs936788471 Chr.16: 23836027-23836027 on Build GRCh38
rs970739626 Chr.16: 23835991-23835991 on Build GRCh38
rs755247718 Chr.16: 23836289-23836289 on Build GRCh38
rs934519852 Chr.16: 23835932-23835932 on Build GRCh38
rs371723231 Chr.16: 23836006-23836006 on Build GRCh38
rs200442462 Chr.16: 23836240-23836240 on Build GRCh38
rs570957353 Chr.16: 23835901-23835901 on Build GRCh38
rs991186247 Chr.16: 23835892-23835892 on Build GRCh38
rs766094053 Chr.16: 23836143-23836143 on Build GRCh38
rs935645982 Chr.16: 23835882-23835882 on Build GRCh38
rs374642745 Chr.16: 23836167-23836167 on Build GRCh38
rs1027154206 Chr.16: 23835987-23835987 on Build GRCh38

View Full Product Details