Assay Design



Gene Transcripts

Gene Symbol
ARNTL
Entrez Gene ID
406
Gene Name
aryl hydrocarbon receptor nuclear translocator like
Gene Aliases
BMAL1, BMAL1c, JAP3, MOP3, PASD3, TIC, bHLHe5
Chromosome Location
Chr.11: 13276613 - 13387268 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030272.2 NP_001025443.1 8 - 9 866 112
NM_001030273.2 NP_001025444.1 9 - 10 937 112
NM_001178.5 NP_001169.3 9 - 10 921 112
NM_001297719.1 NP_001284648.1 9 - 10 921 112
NM_001297722.1 NP_001284651.1 8 - 9 866 112
NM_001297724.1 NP_001284653.1 9 - 10 930 112
XM_006718233.3 XP_006718296.1 8 - 9 808 112
XM_006718234.1 XP_006718297.1 9 - 10 874 112
XM_011520105.2 XP_011518407.2 7 - 8 972 112
XM_011520107.2 XP_011518409.2 7 - 8 967 112
XM_011520108.1 XP_011518410.1 9 - 10 914 112
XM_011520109.1 XP_011518411.1 7 - 8 669 112
XM_011520110.1 XP_011518412.1 10 - 11 929 112
XM_011520111.1 XP_011518413.1 8 - 9 744 112
XM_011520112.2 XP_011518414.1 10 - 11 929 112
XM_011520113.1 XP_011518415.1 10 - 11 929 112
XM_017017738.1 XP_016873227.1 7 - 8 970 112
XM_017017739.1 XP_016873228.1 7 - 8 966 112
XM_017017740.1 XP_016873229.1 8 - 9 808 112
XM_017017741.1 XP_016873230.1 7 - 8 669 112
XM_017017742.1 XP_016873231.1 7 - 8 669 112
XM_017017743.1 XP_016873232.1 7 - 8 669 112
XM_017017744.1 XP_016873233.1 10 - 11 929 112
XM_017017745.1 XP_016873234.1 9 - 10 874 112
XM_017017746.1 XP_016873235.1 9 - 10 884 112
XM_017017747.1 XP_016873236.1 8 - 9 744 112
XM_017017748.1 XP_016873237.1 8 - 9 801 112
GenBank mRNA AB000812.1 - 6 - 7 502 112
AB000815.1 - 7 - 8 573 112
AB000816.1 - 8 - 9 913 112
AF044288.1 - 9 - 10 831 112
AK095749.1 - 9 - 10 822 112
AK291510.1 - 8 - 9 779 112
AK315174.1 - 9 - 10 859 112
BC016674.1 - 6 - 7 465 112
BC031214.1 - 9 - 10 762 112
BC041129.1 - 9 - 10 845 112
D89722.1 - 6 - 7 478 112
U51627.1 - 3 - 4 460 112

View Full Product Details