Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206524.1 | NP_001193453.1 | 1 - 2 | 416 | 65 |
NM_001206542.1 | NP_001193471.1 | 1 - 2 | 416 | 65 | |
NM_001230.4 | NP_001221.2 | 1 - 2 | 416 | 65 | |
NM_001306083.1 | NP_001293012.1 | 1 - 2 | 433 | 65 | |
NM_032974.4 | NP_116756.2 | 1 - 2 | 416 | 65 | |
NM_032976.3 | NP_116758.1 | 1 - 2 | 416 | 65 | |
NM_032977.3 | NP_116759.2 | 1 - 2 | 416 | 65 | |
XM_005246907.2 | XP_005246964.1 | 1 - 2 | 202 | 65 | |
XM_006712796.3 | XP_006712859.1 | 1 - 2 | 429 | 65 | |
XM_011511990.2 | XP_011510292.1 | 1 - 2 | 202 | 65 | |
GenBank mRNA | AJ487678.1 | - | 1 - 2 | 122 | 65 |
AJ487679.1 | - | 1 - 2 | 119 | 65 | |
AK225475.1 | - | 1 - 2 | 154 | 65 | |
AK304859.1 | - | 1 - 2 | 173 | 65 | |
AK310746.1 | - | 1 - 2 | 202 | 65 | |
AY690601.3 | - | 1 - 2 | 154 | 65 | |
BC042844.1 | - | 1 - 2 | 152 | 65 | |
BP871945.1 | - | 1 - 2 | 416 | 65 | |
DA456072.1 | - | 1 - 2 | 434 | 65 | |
U60519.1 | - | 1 - 2 | 145 | 65 | |
U86214.1 | - | 1 - 2 | 192 | 65 |