Assay Design
Gene Transcripts
- Gene Symbol
- EFNA2
- Entrez Gene ID
-
1943
- Gene Name
- ephrin A2
- Gene Aliases
-
ELF-1, EPLG6, HEK7-L, LERK-6, LERK6
- Chromosome Location
-
Chr.19: 1282817 - 1301431 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001405.3 |
NP_001396.2 |
1 - 2 |
158 |
89 |
GenBank mRNA |
AJ007292.1 |
- |
1 - 2 |
158 |
89 |
View Full Product Details