Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127610.1 | NP_001121082.1 | 2 - 3 | 573 | 63 |
NM_001701.3 | NP_001692.1 | 2 - 3 | 674 | 63 | |
GenBank mRNA | AK315347.1 | - | 2 - 3 | 618 | 63 |
BC009567.1 | - | 2 - 3 | 674 | 63 | |
BC107424.1 | - | 2 - 3 | 678 | 63 | |
CR541918.1 | - | 1 - 2 | 464 | 63 | |
L34081.1 | - | 2 - 3 | 648 | 63 |