Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178029.1 | NP_001171500.1 | 6 - 7 | 731 | 66 |
NM_001919.3 | NP_001910.2 | 6 - 7 | 782 | 66 | |
GenBank mRNA | AK291127.1 | - | 6 - 7 | 782 | 66 |
BC000762.2 | - | 6 - 7 | 747 | 66 | |
BC002746.2 | - | 6 - 7 | 702 | 66 | |
BC009631.1 | - | 7 - 8 | 724 | 66 | |
BC019316.1 | - | 6 - 7 | 747 | 66 | |
BC020228.1 | - | 6 - 7 | 702 | 66 | |
BQ773923.1 | - | 1 - 2 | 66 | ||
Z25820.1 | - | 6 - 7 | 755 | 66 |