Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001925.2 | NP_001916.1 | 2 - 3 | 296 | 82 |
GenBank mRNA | BC093959.1 | - | 2 - 3 | 220 | 82 |
BC106747.2 | - | 1 - 2 | 191 | 82 | |
BC112091.1 | - | 2 - 3 | 220 | 82 | |
BU616655.1 | - | 1 - 2 | 82 | ||
DN998607.1 | - | 2 - 3 | 245 | 82 | |
X65977.1 | - | 2 - 3 | 222 | 82 |