Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AF332168.1 | - | 1 - 2 | 42 | 75 |
AK290859.1 | - | 1 - 2 | 80 | 75 | |
AK292813.1 | - | 1 - 2 | 80 | 75 | |
AK303889.1 | - | 1 - 2 | 80 | 75 | |
AY137470.1 | - | 1 - 2 | 42 | 75 | |
AY165046.1 | - | 1 - 2 | 54 | 75 | |
AY178998.2 | - | 1 - 2 | 42 | 75 | |
AY493417.1 | - | 1 - 2 | 152 | 75 | |
AY875691.1 | - | 1 - 2 | 42 | 75 | |
BC015822.1 | - | 1 - 2 | 84 | 75 | |
BC015823.1 | - | 1 - 2 | 86 | 75 | |
BC022347.1 | - | 1 - 2 | 80 | 75 | |
BP342557.1 | - | 1 - 2 | 283 | 75 | |
DQ522304.1 | - | 1 - 2 | 42 | 75 | |
EU779935.1 | - | 1 - 2 | 42 | 75 | |
KF992208.1 | - | 1 - 2 | 54 | 75 | |
KT006756.1 | - | 1 - 2 | 42 | 75 | |
M83202.1 | - | 1 - 2 | 49 | 75 | |
M93150.1 | - | 1 - 2 | 59 | 75 | |
U07643.1 | - | 1 - 2 | 139 | 75 | |
X52941.1 | - | 1 - 2 | 36 | 75 | |
X53961.1 | - | 1 - 2 | 336 | 75 |