Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002677.3 | NP_002668.1 | 3 - 4 | 446 | 77 |
GenBank mRNA | AK307118.1 | - | 2 - 3 | 228 | 77 |
AK311758.1 | - | 3 - 4 | 401 | 77 | |
BC034997.1 | - | 3 - 4 | 451 | 77 | |
CR541649.1 | - | 3 - 4 | 348 | 77 | |
CR541738.1 | - | 3 - 4 | 348 | 77 | |
X62167.1 | - | 3 - 4 | 382 | 77 |