Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003041.3 | NP_003032.1 | 1 - 2 | 143 | 78 |
NR_130783.1 | 1 - 2 | 143 | 78 | ||
XM_006721072.3 | XP_006721135.2 | 1 - 2 | 726 | 78 | |
GenBank mRNA | BC131542.1 | - | 1 - 2 | 143 | 78 |
M95549.1 | - | 1 - 2 | 144 | 78 |