Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166496.1 | NP_001159968.1 | 4 - 5 | 2064 | 110 |
NM_003051.3 | NP_003042.3 | 4 - 5 | 1595 | 110 | |
XM_011542026.2 | XP_011540328.1 | 4 - 5 | 1328 | 110 | |
XM_011542027.2 | XP_011540329.1 | 4 - 5 | 1338 | 110 | |
GenBank mRNA | AK000641.1 | - | 1 - 2 | 480 | 110 |
AK094064.1 | - | 4 - 5 | 2064 | 110 | |
AK296688.1 | - | 4 - 5 | 1378 | 110 | |
AK303603.1 | - | 4 - 5 | 1437 | 110 | |
AK312500.1 | - | 4 - 5 | 1438 | 110 | |
AL162079.1 | - | 4 - 5 | 1425 | 110 | |
BC026317.1 | - | 4 - 5 | 1438 | 110 | |
L31801.1 | - | 3 - 4 | 1244 | 110 |