Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003063.2 | NP_003054.1 | 1 - 2 | 98 | 71 |
GenBank mRNA | AK312097.1 | - | 1 - 2 | 93 | 71 |
BC005261.1 | - | 1 - 2 | 93 | 71 | |
BC094685.1 | - | 1 - 2 | 71 | ||
BC104150.2 | - | 1 - 2 | 50 | 71 | |
BC104185.2 | - | 1 - 2 | 45 | 71 | |
BC113930.1 | - | 1 - 2 | 49 | 71 | |
BP233121.1 | - | 1 - 2 | 71 | ||
U96094.1 | - | 1 - 2 | 71 |