Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198253.2 | NP_937983.2 | 11 - 12 | 2905 | 85 |
XM_017009796.1 | XP_016865285.1 | 11 - 12 | 3477 | 85 | |
GenBank mRNA | AF015950.1 | - | 11 - 12 | 2902 | 85 |
AF018167.1 | - | 11 - 12 | 2903 | 85 | |
AX810036.1 | - | 11 - 12 | 2902 | 85 | |
BC062321.1 | - | 7 - 8 | 1478 | 85 | |
JF896283.1 | - | 4 - 5 | 517 | 85 |