Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003227.3 | NP_003218.2 | 3 - 4 | 516 | 118 |
XM_005250553.4 | XP_005250610.1 | 4 - 5 | 822 | 118 | |
XM_017012573.1 | XP_016868062.1 | 4 - 5 | 624 | 118 | |
GenBank mRNA | AF067864.1 | - | 3 - 4 | 502 | 118 |
BC142630.1 | - | 3 - 4 | 502 | 118 | |
BC150192.1 | - | 3 - 4 | 480 | 118 |