Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003344.3 | NP_003335.1 | 1 - 2 | 459 | 68 |
NM_182697.2 | NP_874356.1 | 1 - 2 | 459 | 68 | |
GenBank mRNA | AB451490.1 | - | 1 - 2 | 54 | 68 |
AK316590.1 | - | 1 - 2 | 42 | 68 | |
AY302138.1 | - | 1 - 2 | 54 | 68 | |
BC006277.2 | - | 1 - 2 | 371 | 68 | |
BT006756.1 | - | 1 - 2 | 54 | 68 | |
CB995681.1 | - | - | 476 | 68 | |
KJ534959.1 | - | 1 - 2 | 54 | 68 | |
Z29328.1 | - | 1 - 2 | 107 | 68 | |
Z29330.1 | - | 1 - 2 | 107 | 68 |