Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000066.3 | NP_000057.2 | 9 - 10 | 1591 | 68 |
NM_001278543.1 | NP_001265472.1 | 10 - 11 | 1813 | 68 | |
NM_001278544.1 | NP_001265473.1 | 10 - 11 | 1720 | 68 | |
XM_017002235.1 | XP_016857724.1 | 10 - 11 | 1659 | 68 | |
GenBank mRNA | AK292740.1 | - | 9 - 10 | 1446 | 68 |
AK298425.1 | - | 10 - 11 | 1637 | 68 | |
AK298444.1 | - | 10 - 11 | 1813 | 68 | |
AK313382.1 | - | 9 - 10 | 1463 | 68 | |
AK316209.1 | - | 10 - 11 | 1810 | 68 | |
BC130575.1 | - | 9 - 10 | 1424 | 68 | |
CD364465.1 | - | 3 - 4 | 68 | ||
M16973.1 | - | 9 - 10 | 1426 | 68 | |
X04393.1 | - | 8 - 9 | 1261 | 68 |