Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000090.3 | NP_000081.1 | 1 - 2 | 200 | 66 |
GenBank mRNA | AK308010.1 | - | 1 - 2 | 200 | 66 |
BC028178.1 | - | 1 - 2 | 198 | 66 | |
BP374999.1 | - | 1 - 2 | 200 | 66 | |
X07240.1 | - | 1 - 2 | 83 | 66 | |
X14420.1 | - | 1 - 2 | 185 | 66 |