Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000164.3 | NP_000155.1 | 7 - 8 | 738 | 116 |
NM_001308418.1 | NP_001295347.1 | 6 - 7 | 630 | 116 | |
XM_011526709.2 | XP_011525011.1 | 7 - 8 | 795 | 116 | |
XM_011526710.2 | XP_011525012.1 | 8 - 9 | 876 | 116 | |
XM_011526711.2 | XP_011525013.1 | 6 - 7 | 687 | 116 | |
XM_011526714.2 | XP_011525016.1 | 4 - 5 | 471 | 116 | |
XM_011526715.2 | XP_011525017.1 | 4 - 5 | 481 | 116 | |
XM_011526716.2 | XP_011525018.1 | 7 - 8 | 795 | 116 | |
GenBank mRNA | BC093723.1 | - | 7 - 8 | 725 | 116 |
BC095494.1 | - | 6 - 7 | 637 | 116 | |
BC101673.1 | - | 7 - 8 | 725 | 116 | |
BC143300.1 | - | 6 - 7 | 618 | 116 | |
BC143302.1 | - | 7 - 8 | 725 | 116 | |
S79852.1 | - | 7 - 8 | 733 | 116 | |
U39231.1 | - | 7 - 8 | 738 | 116 |