Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 28396563-28397070 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SARM1
Entrez Gene ID
23098
Gene Name
sterile alpha and TIR motif containing 1
Gene Aliases
MyD88-5, SAMD2, SARM
UniGene
Hs.743510
Gene Chromosome Location
Chr.17: 28371662-28401045 on Build GRCh38
Transcripts
NM_015077.3 NM_001242366.2 NM_080669.5 XM_005277786.3 XM_017024110.1

Gene Symbol
SLC46A1
Entrez Gene ID
113235
Gene Name
solute carrier family 46 member 1
Gene Aliases
G21, HCP1, PCFT
UniGene
Hs.446689
Gene Chromosome Location
Chr.17: 28394642-28406630 on Build GRCh38
Transcripts
NM_015077.3 NM_001242366.2 NM_080669.5 XM_005277786.3 XM_017024110.1

SNP Detail

SNP ID Chromosome Location
rs764863596 Chr.17: 28397019-28397019 on Build GRCh38
rs988470260 Chr.17: 28396641-28396641 on Build GRCh38
rs2239911 Chr.17: 28396594-28396594 on Build GRCh38
rs576583917 Chr.17: 28396621-28396621 on Build GRCh38
rs901050930 Chr.17: 28396783-28396783 on Build GRCh38
rs998519154 Chr.17: 28396786-28396786 on Build GRCh38
rs980777585 Chr.17: 28396871-28396871 on Build GRCh38
rs551240561 Chr.17: 28396875-28396875 on Build GRCh38
rs955828002 Chr.17: 28396806-28396806 on Build GRCh38
rs12453383 Chr.17: 28396688-28396688 on Build GRCh38
rs1028128966 Chr.17: 28396788-28396788 on Build GRCh38
rs533548408 Chr.17: 28396967-28396967 on Build GRCh38
rs569526091 Chr.17: 28396804-28396804 on Build GRCh38
rs139343092 Chr.17: 28396932-28396932 on Build GRCh38
rs930788238 Chr.17: 28396911-28396911 on Build GRCh38
rs886052745 Chr.17: 28396995-28396995 on Build GRCh38
rs569425471 Chr.17: 28396883-28396883 on Build GRCh38
rs555187371 Chr.17: 28397024-28397024 on Build GRCh38
rs1049158018 Chr.17: 28396739-28396739 on Build GRCh38
rs930719243 Chr.17: 28396716-28396716 on Build GRCh38
rs146914333 Chr.17: 28396814-28396814 on Build GRCh38
rs940758723 Chr.17: 28396955-28396955 on Build GRCh38
rs955692472 Chr.17: 28396605-28396605 on Build GRCh38
rs886052744 Chr.17: 28396848-28396848 on Build GRCh38
rs11871687 Chr.17: 28396910-28396910 on Build GRCh38
rs1004881370 Chr.17: 28396780-28396780 on Build GRCh38
rs1049294138 Chr.17: 28396915-28396915 on Build GRCh38
rs562433674 Chr.17: 28396653-28396653 on Build GRCh38
rs886554110 Chr.17: 28396746-28396746 on Build GRCh38
rs927465028 Chr.17: 28396696-28396696 on Build GRCh38
rs536990699 Chr.17: 28396888-28396888 on Build GRCh38
rs561300818 Chr.17: 28396802-28396802 on Build GRCh38
rs118138669 Chr.17: 28396813-28396813 on Build GRCh38
rs2239910 Chr.17: 28396647-28396647 on Build GRCh38
rs782678539 Chr.17: 28396781-28396781 on Build GRCh38
rs756767 Chr.17: 28396714-28396714 on Build GRCh38
rs739439 Chr.17: 28396803-28396803 on Build GRCh38
rs916907944 Chr.17: 28396829-28396829 on Build GRCh38
rs782382359 Chr.17: 28397020-28397020 on Build GRCh38
rs971642439 Chr.17: 28396836-28396836 on Build GRCh38
rs376935474 Chr.17: 28396759-28396759 on Build GRCh38

View Full Product Details