Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000236.2 | NP_000227.2 | 2 - 3 | 330 | 78 |
XM_005254372.1 | XP_005254429.1 | 3 - 4 | 378 | 78 | |
XM_005254374.4 | XP_005254431.2 | 2 - 3 | 1672 | 78 | |
XM_006720502.3 | XP_006720565.1 | 2 - 3 | 275 | 78 | |
XM_017022176.1 | XP_016877665.1 | 2 - 3 | 1687 | 78 | |
GenBank mRNA | AK290062.1 | - | 4 - 5 | 881 | 78 |
AK292631.1 | - | 4 - 5 | 884 | 78 | |
AK315306.1 | - | 2 - 3 | 509 | 78 | |
BC132825.1 | - | 2 - 3 | 308 | 78 | |
BC136495.1 | - | 2 - 3 | 308 | 78 | |
BC146659.1 | - | 3 - 4 | 553 | 78 | |
D83548.1 | - | 2 - 3 | 314 | 78 | |
J03540.1 | - | 2 - 3 | 277 | 78 | |
J03895.1 | - | 2 - 3 | 280 | 78 | |
X07228.1 | - | 2 - 3 | 330 | 78 |