Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000255.3 | NP_000246.2 | 1 - 2 | 225 | 63 |
GenBank mRNA | AK292568.1 | - | 1 - 2 | 201 | 63 |
AK312611.1 | - | 1 - 2 | 200 | 63 | |
BC016282.1 | - | 1 - 2 | 88 | 63 | |
DA777441.1 | - | 1 - 2 | 231 | 63 | |
DB290266.1 | - | 1 - 2 | 215 | 63 | |
M65131.1 | - | 1 - 2 | 36 | 63 |