Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000324.2 | NP_000315.2 | 7 - 8 | 1133 | 107 |
GenBank mRNA | AB938314.1 | - | 7 - 8 | 1071 | 107 |
AF031548.1 | - | 7 - 8 | 1071 | 107 | |
AF178841.1 | - | 7 - 8 | 1098 | 107 | |
AF179682.1 | - | 7 - 8 | 1098 | 107 | |
AF179684.1 | - | 7 - 8 | 1098 | 107 | |
AF179685.1 | - | 7 - 8 | 1098 | 107 | |
AF187847.1 | - | 7 - 8 | 1098 | 107 | |
AK313505.1 | - | 7 - 8 | 1133 | 107 | |
BC012605.1 | - | 7 - 8 | 1102 | 107 | |
KJ543706.1 | - | 7 - 8 | 1071 | 107 | |
X64594.1 | - | 7 - 8 | 1098 | 107 |