Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000335.4 | NP_000326.2 | 12 - 13 | 2084 | 85 |
NM_001099404.1 | NP_001092874.1 | 12 - 13 | 2084 | 85 | |
NM_001099405.1 | NP_001092875.1 | 12 - 13 | 2084 | 85 | |
NM_001160160.1 | NP_001153632.1 | 12 - 13 | 2085 | 85 | |
NM_001160161.1 | NP_001153633.1 | 12 - 13 | 2085 | 85 | |
NM_198056.2 | NP_932173.1 | 12 - 13 | 2084 | 85 | |
XM_011533991.2 | XP_011532293.1 | 12 - 13 | 2079 | 85 | |
XM_017007017.1 | XP_016862506.1 | 12 - 13 | 2079 | 85 | |
GenBank mRNA | AB158469.2 | - | 11 - 12 | 1932 | 85 |
AB158470.2 | - | 11 - 12 | 1932 | 85 | |
AB208866.1 | - | 3 - 4 | 628 | 85 | |
AF482988.1 | - | 11 - 12 | 1899 | 85 | |
AY038064.1 | - | 11 - 12 | 1948 | 85 | |
AY148488.1 | - | 11 - 12 | 1890 | 85 | |
BC140813.1 | - | 11 - 12 | 1915 | 85 | |
BC144621.1 | - | 11 - 12 | 1915 | 85 | |
EF629346.1 | - | 11 - 12 | 1932 | 85 | |
EF629347.1 | - | 11 - 12 | 1932 | 85 | |
KC858891.1 | - | 11 - 12 | 1890 | 85 | |
M77235.1 | - | 12 - 13 | 2040 | 85 |