Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000444.5 | NP_000435.3 | 11 - 12 | 1867 | 134 |
NM_001282754.1 | NP_001269683.1 | 11 - 12 | 1867 | 134 | |
XM_011545533.1 | XP_011543835.1 | 7 - 8 | 676 | 134 | |
XM_011545536.2 | XP_011543838.1 | 3 - 4 | 709 | 134 | |
XM_017029579.1 | XP_016885068.1 | 7 - 8 | 677 | 134 | |
GenBank mRNA | AD000712.1 | - | 11 - 12 | 1464 | 134 |
AK298030.1 | - | 11 - 12 | 1413 | 134 | |
AK301528.1 | - | 10 - 11 | 1603 | 134 | |
AK304551.1 | - | 4 - 5 | 704 | 134 | |
BC105057.1 | - | 11 - 12 | 1389 | 134 | |
BC105059.1 | - | 11 - 12 | 1389 | 134 | |
U60475.1 | - | 11 - 12 | 1298 | 134 | |
U75645.1 | - | 11 - 12 | 1381 | 134 | |
U82970.1 | - | 11 - 12 | 1908 | 134 | |
U87284.1 | - | 11 - 12 | 1506 | 134 |