Assay Design
Gene Transcripts
- Gene Symbol
- EPX
- Entrez Gene ID
-
8288
- Gene Name
- eosinophil peroxidase
- Gene Aliases
-
EPO, EPP, EPX-PEN, EPXD
- Chromosome Location
-
Chr.17: 58192724 - 58205174 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_000502.5 |
NP_000493.1 |
11 - 12 |
2071 |
84 |
View Full Product Details