Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000545.6 | NP_000536.5 | 2 - 3 | 732 | 96 |
NM_001306179.1 | NP_001293108.1 | 2 - 3 | 732 | 96 | |
XM_005253931.3 | XP_005253988.1 | 2 - 3 | 785 | 96 | |
GenBank mRNA | AK298422.1 | - | 2 - 3 | 606 | 96 |
BC104908.1 | - | 2 - 3 | 551 | 96 | |
BC104910.1 | - | 2 - 3 | 551 | 96 | |
BC143483.1 | - | 2 - 3 | 551 | 96 | |
HM116541.1 | - | 2 - 3 | 674 | 96 | |
HM116542.1 | - | 2 - 3 | 674 | 96 | |
HM116543.1 | - | 2 - 3 | 674 | 96 | |
HM116544.1 | - | 2 - 3 | 674 | 96 | |
HM116547.1 | - | 2 - 3 | 674 | 96 | |
HM116548.1 | - | 2 - 3 | 674 | 96 | |
HM116552.1 | - | 3 - 4 | 291 | 96 | |
HM116558.1 | - | 2 - 3 | 649 | 96 | |
HM449088.1 | - | 2 - 3 | 554 | 96 | |
M57732.1 | - | 2 - 3 | 554 | 96 | |
X71346.1 | - | 2 - 3 | 727 | 96 | |
X71347.1 | - | 2 - 3 | 662 | 96 |