Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291345.1 | NP_001278274.1 | 5 - 6 | 665 | 64 |
NM_001787.2 | NP_001778.2 | 5 - 6 | 695 | 64 | |
NM_033486.2 | NP_277021.2 | 5 - 6 | 695 | 64 | |
NM_033489.2 | NP_277024.2 | 6 - 7 | 742 | 64 | |
XM_006711065.3 | XP_006711128.1 | 5 - 6 | 1407 | 64 | |
XM_017002928.1 | XP_016858417.1 | 6 - 7 | 1484 | 64 | |
GenBank mRNA | AF067512.1 | - | 5 - 6 | 609 | 64 |
AF067514.1 | - | 6 - 7 | 623 | 64 | |
AF067515.1 | - | 5 - 6 | 609 | 64 | |
AF067516.1 | - | 6 - 7 | 656 | 64 | |
AK000081.1 | - | 4 - 5 | 454 | 64 | |
AK304679.1 | - | 5 - 6 | 573 | 64 | |
BC009375.2 | - | 5 - 6 | 606 | 64 | |
BC140714.1 | - | 5 - 6 | 568 | 64 | |
BC171773.1 | - | 5 - 6 | 568 | 64 | |
U04817.1 | - | 5 - 6 | 576 | 64 | |
U04824.1 | - | 5 - 6 | 609 | 64 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001313896.1 | NP_001300825.1 | 5 - 6 | 783 | 64 |
NM_024011.3 | NP_076916.2 | 5 - 6 | 813 | 64 | |
NM_033529.3 | NP_277071.2 | 5 - 6 | 783 | 64 | |
NR_132739.1 | 5 - 6 | 813 | 64 | ||
NR_132740.1 | 5 - 6 | 813 | 64 | ||
GenBank mRNA | AF067518.1 | - | 5 - 6 | 571 | 64 |
AF067519.1 | - | 5 - 6 | 571 | 64 | |
AF067521.1 | - | 5 - 6 | 541 | 64 | |
AF067522.1 | - | 5 - 6 | 541 | 64 | |
AF067523.1 | - | 5 - 6 | 571 | 64 | |
AF067524.1 | - | 2 - 3 | 240 | 64 | |
AK303446.1 | - | 5 - 6 | 571 | 64 | |
AK303451.1 | - | 5 - 6 | 554 | 64 | |
BC028187.1 | - | 5 - 6 | 662 | 64 | |
U07704.1 | - | 5 - 6 | 603 | 64 |