Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001185156.1 | NP_001172085.1 | 3 - 4 | 519 | 83 |
NM_001185157.1 | NP_001172086.1 | 3 - 4 | 519 | 83 | |
NM_006850.3 | NP_006841.1 | 3 - 4 | 516 | 83 | |
XM_011509101.2 | XP_011507403.1 | 2 - 3 | 411 | 83 | |
XM_017000121.1 | XP_016855610.1 | 2 - 3 | 409 | 83 | |
GenBank mRNA | AY641441.1 | - | 3 - 4 | 518 | 83 |
BC009681.1 | - | 3 - 4 | 492 | 83 | |
BP222545.1 | - | 3 - 4 | 516 | 83 | |
BT007156.1 | - | 2 - 3 | 244 | 83 | |
U16261.1 | - | 3 - 4 | 515 | 83 |