Assay Design



Gene Transcripts

Gene Symbol
VTN
Entrez Gene ID
7448
Gene Name
vitronectin
Gene Aliases
V75, VN, VNT
Chromosome Location
Chr.17: 28367277 - 28370352 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000638.3 NP_000629.3 6 - 7 1126 77
GenBank mRNA AK298441.1 - 5 - 6 1175 77
AK312968.1 - 6 - 7 1078 77
BC005046.1 - 6 - 7 1057 77
X03168.1 - 6 - 7 1038 77

View Full Product Details