Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003225.2 | NP_003216.1 | 1 - 2 | 123 | 105 |
GenBank mRNA | BC032811.1 | - | 1 - 2 | 107 | 105 |
BQ083523.1 | - | 1 - 2 | 182 | 105 | |
BU539534.1 | - | - | 105 | ||
X00474.1 | - | 1 - 2 | 123 | 105 | |
X52003.1 | - | 1 - 2 | 123 | 105 |