Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004148.3 | NP_004139.2 | 1 - 2 | 149 | 121 |
GenBank mRNA | BC000298.1 | - | 1 - 2 | 114 | 121 |
BC004440.1 | - | 1 - 2 | 105 | 121 | |
BC019336.1 | - | 1 - 2 | 104 | 121 | |
BC048212.1 | - | 1 - 2 | 149 | 121 | |
BT007164.1 | - | 1 - 2 | 78 | 121 | |
U72661.1 | - | 1 - 2 | 98 | 121 | |
U91512.1 | - | 1 - 2 | 98 | 121 |