Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319108.1 | NP_001306037.1 | 2 - 3 | 399 | 80 |
NM_020530.5 | NP_065391.1 | 2 - 3 | 218 | 80 | |
GenBank mRNA | AB451493.1 | - | 1 - 2 | 99 | 80 |
BC011589.1 | - | 2 - 3 | 215 | 80 | |
BM562358.1 | - | 2 - 3 | 411 | 80 | |
CR456534.1 | - | 2 - 3 | 188 | 80 | |
CR541703.1 | - | 2 - 3 | 174 | 80 | |
CR981101.1 | - | 2 - 3 | 218 | 80 |