Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012329.1 | NP_001012329.1 | 4 - 5 | 461 | 105 |
NM_020248.2 | NP_064633.1 | 5 - 6 | 495 | 105 | |
XM_017001853.1 | XP_016857342.1 | 5 - 6 | 720 | 105 | |
XM_017001854.1 | XP_016857343.1 | 5 - 6 | 594 | 105 | |
XM_017001855.1 | XP_016857344.1 | 4 - 5 | 437 | 105 | |
GenBank mRNA | AB021262.1 | - | 5 - 6 | 461 | 105 |
BC014300.1 | - | 4 - 5 | 449 | 105 | |
BC146892.1 | - | 2 - 3 | 224 | 105 | |
BC146896.1 | - | 2 - 3 | 224 | 105 | |
CB995652.1 | - | - | 488 | 105 |