Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014391.2 | NP_055206.2 | 8 - 9 | 1096 | 63 |
GenBank mRNA | AK290960.1 | - | 8 - 9 | 908 | 63 |
AY903446.1 | - | 8 - 9 | 865 | 63 | |
BC018667.1 | - | 8 - 9 | 912 | 63 | |
X83703.1 | - | 8 - 9 | 1097 | 63 |