Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001303135.1 | NP_001290064.1 | 10 - 11 | 1089 | 86 |
NM_006229.3 | NP_006220.1 | 10 - 11 | 1099 | 86 | |
XM_011539868.1 | XP_011538170.1 | 10 - 11 | 1232 | 86 | |
XM_011539869.1 | XP_011538171.1 | 10 - 11 | 1500 | 86 | |
GenBank mRNA | AK130236.1 | - | 10 - 11 | 1087 | 86 |
BC025784.2 | - | 9 - 10 | 1062 | 86 | |
CD622211.1 | - | 3 - 4 | 86 | ||
CR749299.1 | - | 9 - 10 | 2858 | 86 | |
CR749524.1 | - | 9 - 10 | 3014 | 86 | |
CR749596.1 | - | 9 - 10 | 3012 | 86 | |
M93283.1 | - | 10 - 11 | 1081 | 86 |