Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000212.2 | NP_000203.2 | 10 - 11 | 1714 | 89 |
GenBank mRNA | BC127666.1 | - | 10 - 11 | 1714 | 89 |
BC127667.1 | - | 10 - 11 | 1718 | 89 | |
J02703.1 | - | 10 - 11 | 1714 | 89 | |
KC120777.1 | - | 10 - 11 | 1715 | 89 | |
KC120778.1 | - | 10 - 11 | 1714 | 89 | |
M20311.1 | - | 10 - 11 | 1710 | 89 | |
M35999.1 | - | 10 - 11 | 1712 | 89 | |
U95204.1 | - | 10 - 11 | 1694 | 89 |