Assay Design



Gene Transcripts

Gene Symbol
CCR1
Entrez Gene ID
1230
Gene Name
C-C motif chemokine receptor 1
Gene Aliases
CD191, CKR-1, CKR1, CMKBR1, HM145, MIP1aR, SCYAR1
Chromosome Location
Chr.3: 46201709 - 46208341 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001295.2 NP_001286.1 1 - 2 63 77
GenBank mRNA BC051306.1 - 1 - 2 51 77
BX337412.2 - 1 - 2 62 76
L10918.1 - 1 - 2 54 77

View Full Product Details