Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003897.3 | NP_003888.2 | 1 - 2 | 240 | 98 |
GenBank mRNA | AF083421.1 | - | 1 - 2 | 214 | 98 |
AK222588.1 | - | 1 - 2 | 237 | 98 | |
AK314162.1 | - | 1 - 2 | 241 | 98 | |
BC000844.2 | - | 1 - 2 | 224 | 98 | |
BC005080.1 | - | 1 - 2 | 226 | 98 | |
BM915952.1 | - | 1 - 2 | 245 | 98 | |
BT006703.1 | - | 1 - 2 | 208 | 98 | |
DA893782.1 | - | 1 - 2 | 241 | 98 | |
S81914.1 | - | 1 - 2 | 226 | 98 | |
Y14551.1 | - | 1 - 2 | 219 | 98 |