Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005561.3 | NP_005552.3 | 1 - 2 | 347 | 93 |
GenBank mRNA | AK092398.1 | - | 2 - 3 | 224 | 93 |
AK301584.1 | - | 1 - 2 | 117 | 93 | |
BC021288.2 | - | 1 - 2 | 122 | 93 | |
BC025335.1 | - | 1 - 2 | 260 | 93 | |
BC093044.1 | - | 1 - 2 | 215 | 93 | |
BU849686.1 | - | 1 - 2 | 232 | 93 | |
J04182.1 | - | 1 - 2 | 93 |