Assay Design



Gene Transcripts

Gene Symbol
MS4A1
Entrez Gene ID
931
Gene Name
membrane spanning 4-domains A1
Gene Aliases
B1, Bp35, CD20, CVID5, LEU-16, MS4A2, S7
Chromosome Location
Chr.11: 60455809 - 60470752 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021950.3 NP_068769.2 1 - 2 137 75
GenBank mRNA AK225630.1 - 1 - 2 99 75
AK300025.1 - 1 - 2 95 75
AK310118.1 - 1 - 2 87 75
BC002807.2 - 1 - 2 87 75
X07203.1 - 1 - 2 74 75
X12530.1 - 1 - 2 77 75

View Full Product Details