Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003381.3 | NP_003372.1 | 2 - 3 | 282 | 71 |
NM_194435.2 | NP_919416.1 | 2 - 3 | 282 | 71 | |
XM_005267135.3 | XP_005267192.1 | 2 - 3 | 234 | 71 | |
XM_006715562.3 | XP_006715625.1 | 2 - 3 | 3185 | 71 | |
GenBank mRNA | AK291959.1 | - | 2 - 3 | 234 | 71 |
AK313950.1 | - | 2 - 3 | 282 | 71 | |
BC009794.1 | - | 2 - 3 | 287 | 71 | |
DA914915.1 | - | 2 - 3 | 282 | 71 | |
M36634.1 | - | 2 - 3 | 175 | 71 |